Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Antibody, R&D Systems™
SDP

Catalog No. NB300144SS
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300144SS 0.025 mL
NB300144 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300144SS Supplier R&D Systems Supplier No. NB300144SS

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody has been used in 72 publications

Laminin Polyclonal specifically detects Laminin in Chicken, Chinese Hamster, Cricetulus (Chinese Hamster), Human, Invertebrate, Mammalia, Mouse, Ovine, Pig, Primate, Rabbit, Rat, Sheep samples. It is validated for Block/Neutralize, Flow Cytometry, Gene Knock-Out, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Laminin
Applications Blocking Assay, Flow Cytometry, Gene Knock-Out, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:5000, Flow Cytometry, Immunohistochemistry 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:500 - 1:2000, Immunohistochemistry-Frozen 1:500 - 1:2000, Immunohistochemistry Free-Floating 1:1000 - 1:5000
Formulation 50% PBS, 50% Glycerol with 5mM Sodium Azide
Gene Accession No. P19137
Gene Alias LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha
Gene Symbols LAMA1
Host Species Rabbit
Immunogen Laminin Antibody was made to Laminin 111 isolated from mouse Engelbreth-Holm-Swarm (EHS) sarcoma cells. [UniProt# P19137]
Molecular Weight of Antigen 337 kDa
Purification Method IgG purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 284217
Test Specificity Laminin Antibody is pan-specific and reacts well with all Laminin isoforms tested: Laminin-1 (alpha-1, beta-1, and gamma-1) and Laminin-2 (alpha-2, beta-1, and gamma-1).
Target Species Chicken, Chinese Hamster, Chinese Hamster, Human, Invertebrate, Mammalia, Mouse, Ovine, Pig, Primate, Rabbit, Rat, Sheep
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.