Learn More
Laminin beta-1 Polyclonal Antibody

Description
Laminin beta-1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | Laminin beta-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | LAMB1 |
| Gene Accession No. | P02469, P07942 |
| Gene Alias | C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015 |
| Gene Symbols | LAMB1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.