Learn More
Description
Immunogen sequence: KLVTSMTKQL GDFWTRMEEL RHQARQQGAE AVQAQQLAEG ASEQALSAQE GFERIKQKYA ELKDRLGQSS MLGEQGARIQ SVKTEAEELF GETMEMMDRM KDMELELLRG SQAIMLRSAD LTGLEKRVEQ Binds to an epitope located within the peptide sequence ASEQALSAQEGFERI as determined by overlapping synthetic peptides. Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
Specifications
Specifications
| Antigen | Laminin beta-3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL3353 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LAMB3 |
| Gene Accession No. | Q13751 |
| Gene Alias | AI1A; BM600-125KDA; Epiligrin subunit bata; kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; LAM5; Lamb3; Laminin B1k chain; laminin beta 3; laminin chain; laminin S B3 chain; laminin subunit beta 3; laminin subunit beta-3; laminin, beta 3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); laminin-5 subunit beta; LAMNB1; Nicein subunit beta; nicein, 125kD; nicein, 125kDa; nicein-125k; nicein-125kDa; RP1-272L16.3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
