Learn More
Medchemexpress LLC Lamivudine | 134678-17-4 | 99.9% | 229.26 g/mol | C8H11N3O3S | 10 MG

Supplier: Medchemexpress LLC HYB0250R10MG
Analytical reference standard of lamivudine, an orally active nucleoside reverse transcriptase inhibitor, supplied as a 10 mg pack for laboratory reference, assay calibration, and analytical method development. High purity and characterized identity support reliable quantitative analysis.
- High purity: 99.85% (reported).
- Intended for analytical and research use.
- CAS number: 134678-17-4.
- Molecular formula: C8H11N3O3S.
- Molecular weight: 229.26 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.