Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Reagent Refills
SDP

Catalog No. 03920378 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Sulfide 1
Sulfide 2
Volume (English):
2.02 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Volume (English)
03-920-378 Sulfide 1 2.02 oz.
03-920-379 Sulfide 2 2.02 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-920-378 Supplier LaMotte™ Supplier No. 4633H/DEL

Testing supplies for the discerning scientist

Refills for sulfide tests, potassium iodide, chlorine, ferroine indicators and more

Specifications

Volume (Metric) 60 mL
Storage Requirements 25°C
Type Sulfide 1
Volume (English) 2.02 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.