Learn More
Med Vet International Thermometer, Partial Immersion, -20° to 150° C, 10/Box
Supplier:  Med Vet International 973107BX
Encompass_Preferred
Non-toxic red-filled thermometers can be used in most laboratory applications. Anti-roll device included. 10/Box
Catalog No. 50-304-6895
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.