Learn More
Description
This antibody recognizes an extracellular/luminal epitope of CD107b / LAMP-2, an extensively glycosylated 100-120 kDa widely expressed lysosome-associated protein. This antibody will not cross-react with mouse or rat.
Specifications
Specifications
| Antigen | LAMP2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | H4B4 |
| Concentration | 0.315 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide |
| Gene | LAMP2 |
| Gene Accession No. | P13473 |
| Gene Alias | CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
