Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP2 Mouse anti-Human, Clone: 2D3B9, Proteintech
SDP

Catalog No. 501736733 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6733 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6733 Supplier Proteintech Group Inc Supplier No. 663011IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysosomes are membrane-bound vacuoles which play a critical role in cellular metabolism. These organelles participate in endocytosis, the synthesis and assembly of acid hydrolases, act as sites for digestion of foreign materials and for specialized autolytic cellular processes. Membrane-associated glycoproteins have an important role in normal lysosomal function. Prior to posttranslational modification, Lysosome Associated Membrane Protein 2 (LAMP 2) is an approximately 45 kDa polypeptide. Mature, functional LAMP 2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of approximately 100-110 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail.

Specifications

Antigen LAMP2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D3B9
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LAMP2
Gene Accession No. P13473
Gene Alias CD107b, LAMP 2, LAMP2, LAMPB, LGP110
Gene Symbols LAMP2
Host Species Mouse
Immunogen LAMP2 Fusion Protein Ag0595
Purification Method Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3920
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.