Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMP5 (BAD-LAMP) Monoclonal Antibody (34.2), eBioscience™

Catalog No. p-9090007
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-977-837 2 mg
14-977-880 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-977-837 Supplier Invitrogen™ Supplier No. 14977837
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody 34.2 recognizes human lysosome-associated membrane protein 5 (LAMP5), a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation. This 34.2 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using high or low pH antigen retrieval and can be used at less than or equal to 1 μg/mL. This 34.2 antibody has been tested by western blot analysis of reduced SK-N-SH cell lysates and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Lysosome-associated membrane protein 5 (LAMP5) is a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMP5 (BAD-LAMP)
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 34.2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% Sodium Azide; pH 7.2
Gene LAMP5
Gene Accession No. Q9UJQ1
Gene Alias 3110035N03Rik; 6330527O06Rik; BADLAMP; BAD-LAMP; brain and dendritic cell associated LAMP; brain and dendritic cell-associated LAMP; Brain-associated LAMP-like protein; C20orf103; LAMP family protein C20orf103; LAMP family protein C20orf103 homolog; LAMP5; LAMP-5; lysosomal associated membrane protein family member 5; lysosomal-associated membrane protein family, member 5; lysosome-associated membrane glycoprotein 5; Lysosome-associated membrane protein 5; Protein C20orf103; RGD1306991; UNC-46
Gene Symbols LAMP5
Host Species Rat
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24141
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.