Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMTOR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596186 Supplier Invitrogen™ Supplier No. PA596186
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEPGAGHLDG HRAGSPSLRQ ALCDGSAVMF SSKERGRCTV INFVPLEAPL RSTPRSRQVT EACGGEGRAV PLGSEPEWSV GGMEATLEQH LEDTMKNPSI VGVLCTDSQG LNLGCRGTLS DEHAGVISVL AQQAAKLTSD PTDIPVVCLE SDNGNIMIQK HDGITVAVHK MAS; Positive Samples: U-251MG, Mouse kidney; Cellular Location: Cytoplasm, Lysosome.

HBXIP, also named as XIP, belongs to the HBXIP family. It is originally identified by its interaction with the C-terminus of hepatitis B virus (HBV) X protein (HBx). HBXIP is a regulator of centrosome dynamics and cytokinesis. HBXIP is able to up-regulate S100A4 though activating STAT4 and inducing DNA methylation of PTEN. HBXIP is a cellular 18kd protein and cytoplasm stain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMTOR5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LAMTOR5
Gene Accession No. O43504, Q9D1L9
Gene Alias 1110003H18Rik; HBV X-interacting protein; HBV X-interacting protein homolog; HBx-interacting protein; HBX-interacting protein homolog; HBXIP; hepatitis B virus x interacting protein; Hepatitis B virus X-interacting protein; hepatitis B virus x-interacting protein (9.6kD); hepatitis B virus X-interacting protein homolog; LAMTOR5; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 5; late endosomal/lysosomal adaptor, MAPK and MTOR activator 5; MGC71071; ragulator complex protein LAMTOR5; XIP
Gene Symbols LAMTOR5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human LAMTOR5 (NP_0063932).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10542, 295357, 68576
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.