Learn More
Hardy Diagnostics™ LanaGram™ Rapid Color Test for Bacterial Gram Reaction
Shop All Hardy Diagnostics Products

Description
- This test is not recommended for use with gram-positive cocci, microaerophilic or obligate anaerobic organisms
- LanaGram™ produces a rapid color change in a tube, gram negatives turn yellow, gram positives remain clear
- Ideal for Bacillus, Listeria, Lactobacillus, and non-fermentors, all of which can be difficult to gram stain
Specifications
Specifications
| Disposable | Single-use |
| No. of Reactions | 15 |
| Analysis Time | 5 to 20 min. |
| Form | Liquid |
| Operating Temperature Range | 35°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.