Learn More
Selleck Chemical LLC Ravoxertinib
SDP
Supplier:  Selleck Chemical LLC S755425MG
Encompass_Preferred
Ravoxertinib (GDC-0994) is a potent orally available and highly selective ERK1/2 inhibitor with IC50 of 1 1 nM and 0 3 nM respectively Phase 1
Catalog No. 50-000-04053
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.