Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum aluminum oxide substrate, 10x10x1mm, polished one side, 100 orientation

Catalog No. AA38495KT
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38495KT 1 Ea.
AA38495KU 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38495KT Supplier Thermo Scientific Chemicals Supplier No. 038495KT

100 Orientation | CAS: 12003-65-5 | LaAlO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-65-5
Molecular Formula LaAlO3
MDL Number MFCD00144550

Specifications

Color Brown-Yellow
Physical Form Plate
Quantity 1 Ea.
Odor Odorless
Chemical Name or Material Lanthanum Aluminum Oxide Substrate, Polished Two Sides
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.