Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO)

Catalog No. p-7049201
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1319809 10 g
AA1319818 50 g
AA1319830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1319809 Supplier Thermo Scientific Chemicals Supplier No. 01319809
Only null left
Add to Cart

CAS: 224183-16-8 | Br3La | 378.62 g/mol

Lanthanum(III) bromide is used as a scintillation material in certain applications. It is used in gamma spectroscopy radioisotope detection. It is used as a source of lanthanum in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 224183-16-8
Molecular Formula Br3La
Molecular Weight (g/mol) 378.62
MDL Number MFCD00798151
InChI Key XKUYOJZZLGFZTC-UHFFFAOYSA-K
Synonym lanthanum iii bromide hydrate, acmc-1chtf, lanthanum bromide hydrate, lanthanum bromide-water 1/3/1
PubChem CID 71317322
SMILES [Br-].[Br-].[Br-].[La+3]

Specifications

Color White
Physical Form Crystalline
Quantity 10 g
Solubility Information Soluble in water.
IUPAC Name lanthanum(3+);tribromide;hydrate
Formula Weight 378.62 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) bromide hydrate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 236-896-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.