Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) bromide, ultra dry, 99.9% (REO)

Catalog No. AA4439609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4439609 10 g
AA4439618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4439609 Supplier Thermo Scientific Chemicals Supplier No. 04439609
Add to Cart

CAS: 13536-79-3 | Br3La | 378.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-79-3
Molecular Formula Br3La
Molecular Weight (g/mol) 378.62
MDL Number MFCD00016161
InChI Key XKUYOJZZLGFZTC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) tribromide
SMILES [Br-].[Br-].[Br-].[La+3]

Specifications

Physical Form Lumps
Quantity 10 g
Formula Weight 378.62
Percent Purity 99.9%
Packaging Ampouled under argon
Chemical Name or Material Lanthanum(III) bromide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-896-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.