Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Solstice

Catalog No. 6002403 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
100 g
6 x 100 g
Packaging:
Ampoule
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-024-03 1 Ea. Ampoule
60-023-90 100 g Poly bottle
60-045-978 6 x 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-024-03 Supplier Solstice Supplier No. 320381EA
Add to Cart
Add to Cart

for 1L standard solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710

Specifications

Quantity 1 Ea.
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
UN Number NONH for all modes of transport
Beilstein 3534976
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Formula Weight 58.44g/mol
Packaging Ampoule
Chemical Name or Material Sodium Chloride Concentrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.