Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) fluoride, anhydrous, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer:  Alfa Aesar 1127318

Quantity 50g

 View more versions of this product

Catalog No. AA1127318


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

13709-38-1
99.99% (REO)
195.9
Lanthanum(III) fluoride, anhydrous
Crystals
F3La
MFCD00011073

Specifications

13709-38-1
99.99% (REO)
White crystalline
MFCD00011073
White
Lanthanum(III) fluoride, anhydrous
Crystals
F3La
195.9
Odorless
50g

Safety and Handling

SDS & Safety Documents

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 237-252-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit