Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) perchlorate hexahydrate

Catalog No. AA1164114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1164114 25 g
AA1164122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1164114 Supplier Thermo Scientific Chemicals Supplier No. 01164114
Only null left
Add to Cart

CAS: 14017-46-0 | Cl3H12LaO18 | 545.33 g/mol

Lanthanum(III) perchlorate hexahydrate participates in the preparation of the Schiff base complexes of lanthanide (III) ions. When hydrated lanthanum perchlorate in ethanolic solution is treated with an ethanolic solution of terpyridine, [La(terpy)3](CIO4)3 is deposited as very small crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14017-46-0
Molecular Formula Cl3H12LaO18
Molecular Weight (g/mol) 545.33
MDL Number MFCD00150245
InChI Key DCEUOMQVLPJQAI-UHFFFAOYSA-K
Synonym lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution
PubChem CID 166977
IUPAC Name lanthanum(3+);triperchlorate
SMILES O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 25 g
UN Number UN1481
Solubility Information Easily soluble in water.
Formula Weight 545.35 (437.26 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Lanthanum(III) perchlorate hexahydrate
Hazard Category H272-H315-H319
Hazard Statement GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-835-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.