Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Oxide, Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCL0100100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCL0100100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCL0100100 Supplier Ricca Chemical Company Supplier No. RDCL0100100B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent grade lanthanum oxide 100 g white poly wide mouth bottle.

Chemical Identifiers

CAS 1312-81-8
Molecular Weight (g/mol) 325.81 g/mol
Synonym Lanthanum Trioxide, Lanthanum Oxide

Specifications

Boiling Point 4200°C
Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 100 g
Grade Reagent
Packaging White Poly Wide Mouth Bottle
Decomposition Information Will Not Occur
Density 6.51 g/cm³
Chemical Name or Material Lanthanum Oxide
Show More Show Less
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.