Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Solution, APHA for Metals, Ricca Chemical
SDP

Catalog No. 42854 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
42854 120 mL Natural Poly Bottle
4285-16 500 mL Natural Poly Bottle
4285-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 42854 Supplier Ricca Chemical Company Supplier No. 42854
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 1312-81-8

Specifications

Packaging Natural Poly Bottle
CAS Min % 72.0, 21.35, 4.27
CAS Max % 75.0, 22.22, 4.45
Methods APHA for Metals
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Quantity 120 mL
Chemical Name or Material Lanthanum Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.