Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LanthaScreen™ Elite Tb-anti-HIS Antibody

Catalog No. PV5863
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5863 25 μg
PV5895 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV5863 Supplier Invitrogen™ Supplier No. PV5863
Only null left
Add to Cart
Add to Cart

LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis.

LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity. The Elite Tb-anti-HIS-tag antibody is a specialized antibody with a broader HIS-tag binding performance than our traditional Tb-anti-HIS-tag antibody.

TRUSTED_SUSTAINABILITY

Specifications

Applications Kinase Assay
Concentration 0.46 mg/mL
Conjugate Tb (Terbium)/FITC
Molecular Weight of Antigen 150 kDa
Quantity 25 μg
Regulatory Status RUO
Research Discipline Kinase Biology
Primary or Secondary Primary
Packaging 1 Each
Content And Storage Elite Terbium-labeled anti-HIS antibody in HEPES buffered saline. Store at -20°C.
Form Frozen
Excitation/Emission 340/490, 520 nm
Shipping Condition Dry Ice
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.