Learn More
Lanx Standard Chemical Protective Overgarment Pants
Catalog No. 191300157
Change view
Click to view available options
Color:
Black
Desert Camouflage
Navy
White
Woodland Camouflage
Size:
2X-Large
Large, Long
Large, Regular
Medium, Long
Medium, Regular
Medium, Short
Small, Short
Small, X-Short
X-Large
45 product options available for selection
Product selection table with 45 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-130-0157 Woodland Camouflage Small, Short
19-130-0183 Desert Camouflage Large, Long
19-130-0185 Black Large, Long
19-130-0180 Black Large, Regular
19-130-0184 Navy Large, Long
19-130-0182 Woodland Camouflage Large, Long
19-130-0186 White Large, Long
19-130-0179 Navy Large, Regular
19-130-0181 White Large, Regular
19-130-0177 Woodland Camouflage Large, Regular
19-130-0173 Desert Camouflage Medium, Long
19-130-0174 Navy Medium, Long
19-130-0175 Black Medium, Long
19-130-0176 White Medium, Long
19-130-0172 Woodland Camouflage Medium, Long
19-130-0170 Black Medium, Regular
19-130-0168 Desert Camouflage Medium, Regular
19-130-0169 Navy Medium, Regular
19-130-0165 Black Medium, Short
19-130-0163 Desert Camouflage Medium, Short
19-130-0171 White Medium, Regular
19-130-0164 Navy Medium, Short
19-130-0167 Woodland Camouflage Medium, Regular
19-130-0166 White Medium, Short
19-130-0162 Woodland Camouflage Medium, Short
19-130-0160 Black Small, Short
19-130-0161 White Small, Short
19-130-0158 Desert Camouflage Small, Short
19-130-0155 Black Small, X-Short
19-130-0153 Desert Camouflage Small, X-Short
19-130-0154 Navy Small, X-Short
19-130-0152 Woodland Camouflage Small, X-Short
19-130-0156 White Small, X-Short
19-130-0188 Desert Camouflage X-Large
19-130-0189 Navy X-Large
19-130-0187 Woodland Camouflage X-Large
19-130-0191 White X-Large
19-130-0195 Black 2X-Large
19-130-0193 Desert Camouflage 2X-Large
19-130-0194 Navy 2X-Large
19-130-0196 White 2X-Large
19-130-0192 Woodland Camouflage 2X-Large
19-130-0178 Desert Camouflage Large, Regular
19-130-0159 Navy Small, Short
19-130-0190 Black X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
45 options
This item is not returnable. View return policy
Catalog No. 19-130-0157 Supplier Lanx Supplier No. CPONFRPSSWOOD
Add to Cart
Add to Cart
This item is not returnable. View return policy
Absorbent, durable, air permeable, user comfortable garments
Specifications
Size Small, Short
Material Composite Fabric
Color Woodland Camouflage
Certifications/Compliance ASTM D1777
Size (Metric) Waist 63.5 to 83.82 cm
Item Description 1.5 mm Thick