Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAP1 Monoclonal Antibody (RL13)
GREENER_CHOICE

Catalog No. MA1074 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1074 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1074 Supplier Invitrogen™ Supplier No. MA1074
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-074 detects lamin associated polypeptide (LAP) 1A, 1B & 1C from rat, human, and mouse samples. This antibody does not cross-react with LAP 2. MA1-074 has been successfully used in Western blot, immunofluorescence, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects 3 distinct proteins of molecular weights ~65, ~56 and ~ 51 kDa representing LAP 1A, 1B, and 1C, respectively in rat liver nuclear envelope fractions. Immunofluorescence staining of LAP 1 in rat liver with MA1-074 results in exclusive labeling of the nuclear periphery and exhibits co-localization with lamin staining. The MA1-074 immunogen is pore complex-lamina fraction isolated from rat liver nuclear envelopes.

Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone RL13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TOR1AIP1
Gene Accession No. Q5JTV8, Q5PQX1, Q921T2
Gene Alias lamina-associated polypeptide 1B; Lamina-associated polypeptide 1C; Lamin-associated protein 1B; LAP1; LAP1B; Lap1c; Tor1aip1; torsin 1A interacting protein 1; torsin A interacting protein 1; torsin-1A-interacting protein 1
Gene Symbols TOR1AIP1
Host Species Mouse
Immunogen Pore complex-lamina fraction isolated from rat liver nuclear envelopes.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208263, 246314, 26092
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.