Learn More
Selleck Chemical LLC Dacinostat (LAQ824) 10mg 404951-53-7 NVP-LAQ824
SDP
Supplier:  Selleck Chemical LLC S109510MG
Encompass_Preferred
Dacinostat (LAQ824, NVP-LAQ824) is a novel HDAC inhibitor with IC50 of 32 nM and is known to activate the p21 promoter. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-872-9
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.