Learn More
Enzo Life Sciences LAR (human), (recombinant) (200 U)
SDP
Supplier:  Enzo Life Sciences BMLSE1130200
Encompass_Preferred
Alternative name: Leukocyte common antigen related protein tyrosine phosphatase. Purity: ≥90% (SDS-PAGE). Formulation: Liquid.  In 50mM TRIS-HCl, pH 7.0, containing 100mM sodium chloride, 2mM EDTA, 5mM dithiothreitol, 0.01% Brij 35, and 30% glycerol. Source: Produced in E. coli. Soluble catalytic LAR-D1 domain (350 aa). Long Term Storage: -80°C.
Catalog No. 50-201-1933
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.