Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore™ Large Reagent Bottles, Narrow Mouth, PP
SDPSCI_ED

Catalog No. S111074 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Quantity:
1 Ea.
6 Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
S111074 2 L 1 Ea.
S111075 2 L 6 Pk.
S111076 4 L 1 Ea.
S111077 4 L 6 Pk.
S111078 8 L 1 Ea.
S111079 8 L 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S111074 Supplier United Scientific™ Supplier No. BNMP2L
Add to Cart
Add to Cart

These large capacity polypropylene reagent bottles feature narrow mouths, integrated shoulder loops (except 2 L) and include a lined HDPE cap for a tight seal.

These large capacity reagent bottles feature narrow mouths, integrated shoulder loops (except 2 L) and include a lined HDPE cap for a tight seal.

  • Constructed of polypropylene, the bottles are translucent, pliable, and have excellent strength for extended use.
  • Ideal for packing, shipping and storage purposes.
  • Cap needs to be removed before autoclaving.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
  • Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces.

Specifications

Capacity (Metric) 2 L
Capacity (English) 67.63 oz.
Color Translucent White
Diameter (Metric) Outer Neck 34.5 mm
For Use With (Application) General Lab Use
Material Polypropylene
Quantity 1 Ea.
Sterility Non-sterile
Product Line UniStore
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 240 mm
Product Type Reagent Bottle
Height (English) 9.45 in.
Closure Material HDPE
Closure Size 34.5 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.