Learn More
United Scientific Supplies Large Demonstration Prism Set
SDPSCI_ED
Catalog No. S96415 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Large demonstration prism set
Catalog No.
S96415 Large demonstration prism set
1 options
Catalog No. S96415 Supplier United Scientific Supplies Supplier No. PST620
Add to Cart
Add to Cart
Includes
Semi-circular lens, equilateral prism (3 in. [7.6cm] sides), double convex lens, double concave lens, single convex lens, single concave lens, and activity guide
Oversized set of six lenses and prisms.
  • Designed for mounting on a magnetic chalk or marker board; each piece has a pre-attached magnetic backing
  • Large lens size (8 in. [20.3cm]) ensures that optics experiments are visible from every part of the classroom
  • Set comes in a sturdy wooden storage case
Specifications
Product Type Prism Set, Large Demonstration
For Use With (Application) Designed for mounting on a magnetic chalk or marker board, each piece has a pre attached magnetic backing
For Use With (Equipment) Magnetic chalk or marker board
Includes Semi-circular lens, equilateral prism, double convex lens, double concave lens, single convex lens, single concave lens and activity guide