Learn More
Andwin Scientific LARGE LASER CRYO-TAGS FOR BOT
SDP
Supplier:  Andwin Scientific 91873020
Encompass_Preferred
The Cryo-Babies white labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. The labels measure 3.00 x 2.00 (76mm x 51mm).
Catalog No. 50-821-025
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.