Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARGE Monoclonal Antibody (LARGE-02), PE
GREENER_CHOICE

Catalog No. PIMA528610
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528610 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528610 Supplier Invitrogen™ Supplier No. MA528610
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes human LARGE1, a glycosyltransferase expressed mainly in the Golgi apparatus. Cross reactivity with LARGE2 was not determined.

The LARGE gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARGE
Applications Flow Cytometry
Classification Monoclonal
Clone LARGE-02
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene Large1
Gene Accession No. O95461
Gene Alias Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Beta-1,3-glucuronyltransferase LARGE; BPFD#36; enr; eosinophil granule major basic protein 1; fg; froggy; glycosyltransferase-like 1A; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; glycosyltransferase-like protein LARGE1; LARGE xylosyl- and glucuronyltransferase 1; Gyltl1a; KIAA0609; large; LARGE xylosyl- and glucuronyltransferase 1; Large1; like-acetylglucosaminyltransferase; like-glycosyltransferase; MBP1; Mbp-1; MDC1D; MDDGA6; MDDGB6; mKIAA0609; myd; myodystrophy; prepro major basic protein 1; Xylosyltransferase LARGE
Gene Symbols Large1
Host Species Mouse
Immunogen Human LARGE1 (amino acids 35-142).
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9215
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.