Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Large Motion Cart
SDPSCI_ED

Catalog No. S000870 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000870 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S000870 Supplier United Scientific™ Supplier No. LGMTNCART
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

This large motion cart is designed to explore and test different motion topics.

The cart body is 12 in. long and 2 in. wide, providing ample experimentation space. Friction-reducing bearings allow the plastic wheels to roll smoothly. The front of the cart has a hole for attaching a string, which can be used to pull an object or connect a falling mass.

  • Rugged design
  • Smooth rolling wheels

Specifications

Grade 5+
Dimensions (L x W) 12 x 12 in.
Material Aluminum
Class Size 1 Lab Group
Product Type Physics Cart
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.