Learn More
United Scientific Supplies Large U Shaped Electromagnet
SDPSCI_ED
Catalog No. S82874 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
Catalog No. Grade
S82874 5+
1 options
This item is not returnable. View return policy
Prop 65 Warning
Catalog No. S82874 Supplier United Scientific Supplies Supplier No. EMGT01
Add to Cart
Add to Cart
This item is not returnable. View return policy
Prop 65 Warning
Large U-Shaped Electromagnet is designed to last in the classroom.
  • Operating on 1.5 to 6 V DC, this electromagnet lifts up to 3.5 kg.
  • A current direction indicator is included for the study of the magnetic field.
Specifications
Shape U-shaped
Grade 5+
Product Type Electromagnet