Learn More
Selleck Chemical LLC Larotrectinib (LOXO-101) sulfate 5mg 1223405-08-0 ARRY-470, Larotrectinib
SDP
Supplier:  Selleck Chemical LLC S79605MG
Encompass_Preferred
Larotrectinib sulfate (LOXO-101, ARRY-470) is an oral potent and selective ATP-competitive inhibitor of tropomyosin receptor kinases (TRK). Larotrectinib inhibition of TRKs induces cellular apoptosis and G1 cell-cycle arrest. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6404
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.