Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554182 Supplier Invitrogen™ Supplier No. PA554182
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLFPNENLPS KMLLVYDLYL SPKLWALATP QKNGRVQEKV MEHLLKLFGT FGVISSVRIL KPGRELPPDI RRISSRYSQV GTQECAIVEF EEVEAAIKAH EFMITESQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.

LARP6 is a member of the La ribonucleoprotein domain family, which all contain one characteristic La motif. LARP6 shuttles between the cytoplasm and nucleus. It is proposed that members of this family play a role in translation and mRNA stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARP6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LARP6
Gene Accession No. Q9BRS8
Gene Alias 5430431G03Rik; acheron; Achn; AI552438; death-associated LA motif protein; La ribonucleoprotein 6, translational regulator; La ribonucleoprotein domain family member 6; La ribonucleoprotein domain family, member 6; la-related protein 6; Larp6; RGD1308414
Gene Symbols LARP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LARP6. Recombinant protein control fragment (Product #RP-91858).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55323
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.