Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassette- Embedding in Quickload™ Stack (Taped)

Catalog No. 22138891 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-138-891 Gray
22-138-885 Blue
22-138-881 Green
22-138-873 Lilac
22-138-875 Orange
22-138-887 Peach
22-138-879 Pink
22-138-889 Tan
22-138-877 White
22-138-883 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-138-891 Supplier Simport Scientific Supplier No. M4809AT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassettes with additive in Quickload™ taped stacks combine permanent laser marking with automated printer compatibility, manufactured from durable acetal, providing reliable specimen identification and excellent resistance to histological solvents

Histosette™ Embedding Laser Cassettes with additive in Quickload™ taped stacks deliver the same reliable performance as our standard embedding cassettes while providing the added convenience of automated cassette feeding. Designed for compatibility with most cassette printers, they help improve workflow efficiency and consistency in high-volume histology laboratories.

Features & Benefits

  • Supplied in convenient Quickload™ taped stacks for automated cassette feeding
  • Compatible with most cassette printers
  • Designed for routine tissue processing, embedding, sectioning, and archiving
  • High-resolution laser marking provides permanent, easy-to-read specimen identification
  • Resistant to commonly used histological solvents, dyes, decalcifying agents, and processing reagents
  • Precision-molded acetal construction offers excellent durability and dimensional stability
  • 45° anterior writing surface optimizes laser printing quality
  • Accepts standard stainless steel lids (M481, sold separately)
  • Available in 10 standard colors
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Recommended for laboratories using automated laser cassette printers that require high-throughput specimen identification while maintaining excellent chemical resistance, permanent laser marking, and dependable tissue protection throughout the histology workflow.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Tissue Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.