Learn More
Description
Histosette™ Embedding Laser Cassettes with additive are engineered to provide reliable specimen identification and secure tissue handling throughout histology workflows. Manufactured from durable acetal, these embedding cassettes combine exceptional chemical resistance with permanent, high-resolution laser marking for accurate specimen traceability from processing to long-term storage.
Features & Benefits
- Designed for routine tissue processing, embedding, sectioning, and archiving
- High-resolution laser marking provides permanent, easy-to-read specimen identification
- Resistant to commonly used histological solvents, dyes, decalcifying agents, and processing reagents
- Precision-molded acetal construction offers excellent durability and dimensional stability
- 45° anterior writing surface improves label readability and printing quality
- Accepts standard stainless steel lids (M481, sold separately)
- Fits securely in standard microtome chuck adapters
- Available in 10 standard colors for efficient laboratory organization
- Produced under an ISO 9001-certified quality management system
- CE-IVD compliant
Applications
Ideal for hospital pathology laboratories, clinical laboratories, research facilities, and histology laboratories requiring reliable specimen identification, excellent chemical resistance, and consistent performance throughout tissue processing and embedding procedures.
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Gray |
| Material | Acetal |
| Product Type | Tissue Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.