Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassette- Histosette™ Biopsy in Quickload™ Stack (Taped)

Catalog No. 22138911 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-138-911 Gray
22-138-908 Blue
22-138-906 Green
22-138-902 Lilac
22-138-903 Orange
22-138-909 Peach
22-138-905 Pink
22-138-910 Tan
22-138-904 White
22-138-907 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-138-911 Supplier Simport Scientific Supplier No. M4939AT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassette with additive in Quickload™ taped stacks combine permanent high-resolution laser marking with automated cassette feeding, designed for small biopsy specimens, improve workflow efficiency while ensuring secure specimen handling

Histosette™ Biopsy Laser Cassettes with additive in Quickload™ taped stacks provide the same patented biopsy cassette design as the standard version while offering the added convenience of automated cassette feeding. Compatible with most cassette printers, they deliver permanent specimen identification, efficient laboratory workflows, and secure handling of delicate biopsy specimens.

Features & Benefits

  • Supplied in convenient Quickload™ taped stacks for automated cassette feeding
  • Compatible with most cassette printers
  • Patented Histosette™ Biopsy design
  • Integrated additive optimized for permanent, high-resolution laser marking
  • Specifically designed for small biopsy specimens
  • 1 mm square openings maximize reagent exchange while securely retaining delicate tissue samples
  • Secure locking mechanism helps protect specimens throughout processing
  • Pre-attached lid with front hinge enables convenient single-handed operation
  • Unobstructed 45° anterior writing surface improves laser printing quality and readability
  • Resistant to commonly used histological solvents, dyes, decalcifying agents, and processing reagents
  • Precision-molded acetal construction provides excellent durability and dimensional stability
  • Packaged in stacks of 40, with 50 stacks per case for a total of 2,000 cassettes
  • Available in 10 standard colors
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Recommended for laboratories using automated cassette printers that require efficient processing of small biopsy specimens while maintaining secure specimen containment, permanent laser identification, and consistent high-throughput workflow performance.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.