Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassette- Histosette™ Biopsy

Catalog No. 21235740 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-235-740 Gray
21-235-737 Blue
21-235-735 Green
21-235-731 Lilac
21-235-732 Orange
21-235-738 Peach
21-235-734 Pink
21-235-739 Tan
21-235-733 White
21-235-736 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 21-235-740 Supplier Simport Scientific Supplier No. M4939A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassette with additive are specifically designed for secure handling of small biopsy specimens during tissue processing, patented design features 1 mm openings, permanent high-resolution laser marking, and excellent resistance to histological solvents

Histosette™ Biopsy Laser Cassettes with additive are specifically engineered for the secure handling and identification of small biopsy specimens throughout histology workflows. Their patented design combines permanent, high-resolution laser marking with 1 mm square openings, providing excellent specimen protection while maximizing reagent exchange during tissue processing.

Features & Benefits

  • Compatible with most cassette printers
  • Patented Histosette™ Biopsy design
  • Integrated additive optimized for permanent, high-resolution laser marking
  • Specifically designed for small biopsy specimens
  • 1 mm square openings maximize reagent exchange while securely retaining delicate tissue samples
  • Secure locking mechanism helps protect specimens throughout processing
  • Pre-attached lid eliminates the need for separate assembly
  • Front hinge design enables convenient single-handed operation
  • Unobstructed 45° anterior writing surface improves laser printing quality and readability
  • Resistant to commonly used histological solvents, dyes, decalcifying agents, and processing reagents
  • Precision-molded acetal construction provides excellent durability and dimensional stability
  • Available in 10 standard colors
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Ideal for hospital pathology laboratories, clinical laboratories, and research facilities processing small biopsy specimens requiring secure containment, permanent laser identification, and reliable reagent penetration throughout routine histology procedures.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.