Learn More
Simport Scientific Laser Cassette Histosette™ Tissue in Quickload™ Stack (Taped)
Shop All Simport Scientific ProductsDescription
Histosette™ Tissue Laser Cassettes with additive in Quickload™ taped stacks deliver reliable specimen handling and permanent identification while providing the added convenience of automated cassette feeding. Manufactured from durable acetal, these cassettes provide high-resolution laser marking and excellent resistance to commonly used histological solvents and reagents.
Features & Benefits
- Supplied in convenient Quickload™ taped stacks for automated cassette feeding
- Compatible with most cassette printers
- Designed for routine tissue processing, embedding, sectioning, and archiving
- High-resolution laser marking provides permanent, easy-to-read specimen identification
- Efficient flow-through openings maximize fluid exchange and promote proper drainage during processing
- Precision-molded acetal construction provides excellent durability and chemical resistance
- Pre-attached lid with front hinge design allows convenient single-handed cassette manipulation
- Secure locking device helps keep specimens safely contained during processing
- 45° unobstructed anterior writing surface optimizes laser printing quality
- Packaged in stacks of 40, with 50 stacks per case for a total of 2,000 cassettes
- Available in 10 standard colors for efficient laboratory organization
- Produced under an ISO 9001-certified quality management system
- CE-IVD compliant
Applications
Ideal for hospital pathology laboratories, clinical laboratories, research facilities, and high-volume histology laboratories using automated cassette printers and requiring efficient cassette feeding, reliable tissue specimen handling, permanent laser identification, and consistent performance throughout routine histology procedures.
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Gray |
| Material | Acetal |
| For Use With (Equipment) | Most cassette printers |
| Product Type | Tissue Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.