Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassette Histosette™ Tissue

Catalog No. 21235730 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-235-730 Gray
21-235-727 Blue
21-235-725 Green
21-235-721 Lilac
21-235-722 Orange
21-235-728 Peach
21-235-724 Pink
21-235-729 Tan
21-235-723 White
21-235-726 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 21-235-730 Supplier Simport Scientific Supplier No. M4929A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassettes with additive are designed for secure specimen identification during routine tissue processing, patented design features a pre-attached lid, front hinge, and permanent high-resolution laser marking with excellent resistance to histological solvents

Histosette™ Tissue Laser Cassettes with additive are designed for secure specimen handling and reliable identification throughout routine histology workflows. Manufactured from durable acetal, these cassettes provide permanent, high-resolution laser marking and excellent resistance to commonly used histological solvents and reagents.

Features & Benefits

  • Compatible with most cassette printers
  • Designed for routine tissue processing, embedding, sectioning, and archiving
  • High-resolution laser marking provides permanent, easy-to-read specimen identification
  • Efficient flow-through openings maximize fluid exchange and promote proper drainage during processing
  • Precision-molded acetal construction provides excellent durability and chemical resistance
  • Pre-attached lid with front hinge design allows convenient single-handed cassette manipulation
  • Secure locking device helps keep specimens safely contained during processing
  • 45° unobstructed anterior writing surface optimizes laser printing quality
  • Available in 10 standard colors for efficient laboratory organization
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Ideal for hospital pathology laboratories, clinical laboratories, research facilities, and histology laboratories requiring reliable tissue specimen handling, permanent laser identification, efficient fluid exchange, and consistent performance throughout routine histology procedures.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Tissue Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.