Learn More
Description
Microsette™ Micro-Biopsy Laser Cassettes with additive are specifically engineered for the secure handling and identification of extremely small biopsy specimens throughout histology workflows. Their patented design eliminates the need for biopsy pads while providing permanent, high-resolution laser marking and exceptional specimen retention. Featuring over 2,000 precision-molded 0.26 mm square openings, these cassettes maximize reagent exchange and ensure efficient tissue processing without compromising specimen security.
Features & Benefits
- Compatible with most cassette printers
- Patented Microsette™ Micro-Biopsy design
- Integrated additive optimized for permanent, high-resolution laser marking
- Specifically designed for extremely small biopsy specimens
- Eliminates the need for biopsy pads
- Over 2,000 precision-molded 0.26 mm square openings maximize reagent exchange while securely retaining delicate tissue samples
- Secure locking mechanism helps protect specimens throughout processing
- Pre-attached lid eliminates the need for separate assembly
- Unobstructed 45° anterior writing surface improves laser printing quality and readability
- Resistant to commonly used histological solvents, dyes, decalcifying agents, and processing reagents
- Precision-molded acetal construction provides excellent durability and dimensional stability
- Available in 10 standard colors
- Produced under an ISO 9001-certified quality management system
- CE-IVD compliant
Applications
Ideal for hospital pathology laboratories, clinical laboratories, and research facilities processing needle biopsies and other extremely small tissue specimens requiring maximum specimen retention, efficient reagent penetration, and permanent laser identification throughout routine histology procedures.
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Gray |
| Material | Acetal |
| For Use With (Equipment) | Most cassette printers |
| Product Type | Micro-biopsy Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.