Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassette- Micromesh™ Biopsy

Catalog No. 21235769 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-235-769 Gray
21-235-763 Blue
21-235-759 Green
21-235-751 Lilac
21-235-753 Orange
21-235-765 Peach
21-235-757 Pink
21-235-767 Tan
21-235-755 White
21-235-761 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 21-235-769 Supplier Simport Scientific Supplier No. M5079A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassettes with additive are designed for secure handling of small biopsy specimens without biopsy pads, featuring 1,676 square 0.38 mm openings, provide excellent specimen retention, rapid fluid exchange, and permanent high-resolution laser identification

MicroMesh™ Biopsy Laser Cassettes with additive are engineered for the secure handling and identification of small biopsy specimens throughout histology workflows. Their unique design eliminates the need for biopsy pads while providing permanent, high-resolution laser marking. Featuring 1,676 precision-molded 0.38 mm square openings, these cassettes provide excellent specimen retention while maximizing fluid exchange and drainage during tissue processing.

Features & Benefits

  • Compatible with most cassette printers
  • Integrated additive optimized for permanent, high-resolution laser marking
  • Specifically designed for small biopsy specimens
  • Eliminates the need for biopsy pads
  • 1,676 precision-molded 0.38 mm square openings maximize fluid exchange and drainage while securely retaining specimens
  • Large anterior slots in both the cassette and cover promote rapid fluid exchange and drainage
  • Large 27 mm² interior compartment provides ample space for needle biopsies
  • Recessed cover does not protrude above the cassette, allowing efficient stacking in automated cassette printers and tissue processors
  • Easy-to-remove pre-attached disposable cover
  • 45° anterior writing surface is suitable for automated cassette printing
  • Resistant to commonly used histological solvents, dyes, and processing reagents
  • Precision-molded acetal construction provides durability throughout histology processing
  • Available in multiple colors
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Ideal for hospital pathology laboratories, clinical laboratories, and research facilities processing small biopsies and needle biopsy specimens that require secure specimen retention, rapid reagent exchange, efficient drainage, and permanent laser identification throughout routine histology procedures.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.