Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassette- Slimsette™ Biopsy in Quickload™ Stack (Taped)

Catalog No. 22138941 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-138-941 Gray
22-138-938 Blue
22-138-936 Green
22-138-932 Lilac
22-138-933 Orange
22-138-939 Peach
22-138-935 Pink
22-138-940 Tan
22-138-934 White
22-138-937 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-138-941 Supplier Simport Scientific Supplier No. M5109AT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassettes with additive in Quickload™ taped stacks combine secure biopsy specimen handling with permanent high-resolution laser identification and efficient automated feeding, 392 square 1 x 1 mm openings promote effective fluid exchange and drainage

Slimsette™ Biopsy Laser Cassettes with additive in Quickload™ taped stacks combine reliable biopsy specimen identification and secure tissue handling with the convenience of automated cassette feeding. Compatible with most cassette printers, they provide permanent, high-resolution laser marking and excellent resistance to commonly used histological solvents. Their space-saving Slimsette™ design features a recessed cover and 392 square 1 x 1 mm openings for efficient fluid exchange and drainage during tissue processing.

Features & Benefits

  • Supplied in convenient Quickload™ taped stacks for automated cassette feeding
  • Compatible with most cassette printers
  • Integrated additive optimized for permanent, high-resolution laser marking
  • Specifically designed for biopsy specimens
  • 392 square openings measuring 1 x 1 mm promote efficient fluid exchange and drainage during tissue processing
  • Recessed cover does not protrude above the cassette, providing a space-saving design for efficient stacking in automated labeling systems and storage cabinets
  • Pre-attached, easy-to-remove disposable cover simplifies cassette handling
  • 45° unobstructed anterior writing surface supports clear and reliable laser identification
  • Resistant to commonly used histological solvents, dyes, and processing reagents
  • Precision-molded acetal construction provides durability throughout histology processing
  • Available in multiple colors
  • Packaged in stacks of 40, with 50 stacks per case for a total of 2,000 cassettes
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Recommended for pathology, clinical, and research laboratories using automated cassette printers to process biopsy specimens. The Quickload™ taped format supports efficient high-throughput workflows while maintaining secure specimen handling, effective reagent exchange and drainage, and permanent laser identification.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.