Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassette- Slimsette™ Tissue

Catalog No. 22138930 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-138-930 Gray
22-138-924 Blue
22-138-920 Green
22-138-912 Lilac
22-138-914 Orange
22-138-926 Peach
22-138-918 Pink
22-138-928 Tan
22-138-916 White
22-138-922 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-138-930 Supplier Simport Scientific Supplier No. M5099A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassettes with additive provide secure tissue handling and permanent high-resolution laser identification, 1,140 openings maximize fluid exchange and drainage, while the recessed cover enables space-saving stacking in automated labeling and storage systems

Slimsette™ Tissue Laser Cassettes with additive are engineered for reliable specimen identification and secure tissue handling throughout histology workflows. Compatible with most cassette printers, they provide permanent, high-resolution laser marking. Their space-saving design features a recessed cover that does not protrude above the cassette, allowing more cassettes to be stacked in automated labeling systems and storage cabinets.

Features & Benefits

  • Compatible with most cassette printers
  • Integrated additive optimized for permanent, high-resolution laser marking
  • Designed for routine tissue processing, embedding, sectioning, and archiving
  • 1,140 openings measuring 1 x 5 mm maximize fluid exchange and promote efficient reagent drainage
  • Recessed cover does not protrude above the cassette, providing a space-saving design for efficient stacking
  • Pre-attached, easy-to-remove disposable cover simplifies cassette handling
  • 45° unobstructed anterior writing surface supports clear and reliable laser identification
  • Resistant to commonly used histological solvents, dyes, and processing reagents
  • Precision-molded acetal construction provides durability throughout histology processing
  • Available in multiple colors
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Ideal for hospital pathology laboratories, clinical laboratories, and research facilities requiring reliable tissue specimen handling, efficient reagent exchange and drainage, permanent laser identification, and space-efficient cassette stacking throughout routine histology workflows.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Tissue Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.