Learn More
Simport Scientific Laser Cassette- Swingsette™ Biopsy in Quickload™ Stacks (Taped)
Shop All Simport Scientific ProductsDescription
Swingsette™ Biopsy Laser Cassettes with additive in Quickload™ taped stacks are specifically designed for secure biopsy specimen handling while providing the convenience of automated cassette feeding. Compatible with most cassette printers, they provide permanent, high-resolution laser marking for reliable specimen identification throughout histology workflows. Molded from durable acetal, the cassettes feature 1 mm square openings that promote efficient fluid exchange and drainage while securely retaining biopsy specimens.
Features & Benefits
- Supplied in convenient Quickload™ taped stacks for automated cassette feeding
- Compatible with most cassette printers
- Integrated additive optimized for permanent, high-resolution laser marking
- Specifically designed for biopsy specimen processing
- 1 mm square openings promote efficient fluid exchange and drainage
- Unique hinged design keeps the cassette base and cover securely connected
- Hinge configuration allows the cover to be opened and closed multiple times
- Large tab facilitates convenient opening and handling
- 45° anterior writing surface supports clear and reliable laser identification
- Resistant to commonly used histological solvents, dyes, and processing reagents
- Precision-molded acetal construction provides durability throughout histology processing
- Covers are packaged separately
- Available in multiple colors
- Packaged in Quickload™ taped stacks of 40 cassettes; 50 stacks per case for a total of 2,000 cassettes
- Produced under an ISO 9001-certified quality management system
- CE-IVD compliant
Applications
Recommended for histology and pathology laboratories using automated cassette printers to process biopsy specimens. The Quickload™ taped format supports efficient high-throughput workflows while the 1 mm square openings promote effective reagent exchange and drainage, and the Swingsette™ hinged design provides convenient specimen handling throughout tissue processing.
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Gray |
| Material | Acetal |
| For Use With (Equipment) | Most cassette printers |
| Product Type | Biopsy Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.