Learn More
Simport Scientific Laser Cassette- Swingsette™ Tissue in Quickload™ Stack (Taped)
Shop All Simport Scientific ProductsDescription
Swingsette™ Tissue Laser Cassettes with additive in Quickload™ taped stacks combine reliable tissue handling with the convenience of automated cassette feeding. Compatible with most cassette printers, they provide permanent, high-resolution laser marking for clear specimen identification and efficient laboratory workflows. Their unique hinged design securely connects the cassette base and cover while large open slots promote efficient fluid exchange and drainage throughout tissue processing.
Features & Benefits
- Supplied in convenient Quickload™ taped stacks for automated cassette feeding
- Compatible with most cassette printers
- Integrated additive optimized for permanent, high-resolution laser marking
- Designed for routine tissue processing, embedding, sectioning, and archiving
- Large open slots promote efficient fluid exchange and drainage
- Unique hinged design keeps the cassette base and cover securely connected
- Hinge configuration allows the cover to be opened and closed multiple times
- Large tab facilitates convenient opening and handling
- 45° anterior writing surface supports clear and reliable laser identification
- Resistant to commonly used histological solvents, dyes, and processing reagents
- Precision-molded acetal construction provides durability throughout histology processing
- Covers are packaged separately
- Available in multiple colors
- Packaged in Quickload™ taped stacks of 40 cassettes; 50 stacks per case for a total of 2,000 cassettes
- Produced under an ISO 9001-certified quality management system
- CE-IVD compliant
Applications
Recommended for histology laboratories using automated cassette printers that require efficient cassette feeding, reliable tissue handling, permanent laser identification, and effective reagent exchange throughout routine tissue processing.
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Gray |
| Material | Acetal |
| For Use With (Equipment) | Most cassette printers |
| Product Type | Tissue Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.