Learn More
Description
Swingsette™ Tissue Laser Cassettes with additive are designed for reliable specimen identification and secure tissue handling throughout histology workflows. Compatible with most cassette printers, they provide permanent, high-resolution laser marking for clear specimen traceability. Molded from durable acetal, the cassettes feature large open slots that promote efficient fluid exchange and drainage during tissue processing. Their unique hinged design securely connects the cassette base and cover while allowing the cover to be opened and closed multiple times.
Features & Benefits
- Compatible with most cassette printers
- Integrated additive optimized for permanent, high-resolution laser marking
- Designed for routine tissue processing, embedding, sectioning, and archiving
- Large open slots promote efficient fluid exchange and drainage
- Unique hinged design keeps the cassette base and cover securely connected
- Hinge configuration allows the cover to be opened and closed multiple times
- Large tab facilitates convenient opening and handling
- 45° anterior writing surface supports clear and reliable laser identification
- Resistant to commonly used histological solvents, dyes, and processing reagents
- Precision-molded acetal construction provides durability throughout histology processing
- Covers are packaged separately
- Available in multiple colors
- Cassette bases are packaged in two dispenser boxes of 500; one dispenser box contains 1,000 covers
- CE-IVD compliant
Applications
Recommended for histology laboratories requiring reliable tissue specimen processing and permanent laser identification. The Swingsette™ hinged design provides convenient specimen access while its large openings promote effective reagent exchange and drainage throughout routine tissue processing.
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Gray |
| Material | Acetal |
| For Use With (Equipment) | Most cassette printers |
| Product Type | Tissue Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.