Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Scientific Laser Cassettes- Slimsette™ Biopsy

Catalog No. 21235780 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-235-780 Gray
21-235-777 Blue
21-235-775 Green
21-235-771 Lilac
21-235-772 Orange
21-235-778 Peach
21-235-774 Pink
21-235-779 Tan
21-235-773 White
21-235-776 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 21-235-780 Supplier Simport Scientific Supplier No. M5109A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cassettes with additive are designed for secure biopsy specimen handling and permanent high-resolution laser identificatio, featuring 392 square 1 x 1 mm openings, provide efficient fluid exchange and drainage and are compatible with most cassette printers

Slimsette™ Biopsy Laser Cassettes with additive are engineered for reliable identification and secure handling of biopsy specimens throughout histology workflows. Compatible with most cassette printers, they provide permanent, high-resolution laser marking and excellent resistance to commonly used histological solvents. Their space-saving Slimsette™ design features a recessed cover and 392 square 1 x 1 mm openings for efficient fluid exchange and drainage during tissue processing.

Features & Benefits

  • Compatible with most cassette printers
  • Integrated additive optimized for permanent, high-resolution laser marking
  • Specifically designed for biopsy specimens
  • 392 square openings measuring 1 x 1 mm promote efficient fluid exchange and drainage during tissue processing
  • Recessed cover does not protrude above the cassette, providing a space-saving design for efficient stacking in automated labeling systems and storage cabinets
  • Pre-attached, easy-to-remove disposable cover simplifies cassette handling
  • 45° unobstructed anterior writing surface supports clear and reliable laser identification
  • Resistant to commonly used histological solvents, dyes, and processing reagents
  • Precision-molded acetal construction provides durability throughout histology processing
  • Available in multiple colors
  • Produced under an ISO 9001-certified quality management system
  • CE-IVD compliant

Applications

Ideal for hospital pathology laboratories, clinical laboratories, and research facilities processing biopsy specimens that require secure specimen handling, efficient reagent exchange and drainage, permanent laser identification, and compatibility with automated cassette printing workflows.

Specifications

Writing Face Angle 45°
Color Gray
Material Acetal
For Use With (Equipment) Most cassette printers
Product Type Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.