Learn More
United Scientific Supplies Laser Diffraction Kit and Accessories
SDPSCI_ED
Catalog No. S27832 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Kit, Laser Diffraction Complete
Laser Pointer Holder
Momentary Toggle Clip
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S27832 Momentary Toggle Clip
S27823 Kit, Laser Diffraction Complete
S27828 Laser Pointer Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S27832 Supplier United Scientific Supplies Supplier No. MTCLIP
Add to Cart
Add to Cart
Allows students create, view, and measure diffraction and interference patterns
  • Explore the mathematics behind diffraction and interference patterns
  • Sturdy base securely holds included red and green laser pointers
  • Project various patterns against bare walls
  • Includes Diffraction Grating (1), Single-Slit Slide (1), Double-Slit Slides (2), Activity Guide
Specifications
Product Type Momentary Toggle Clip
For Use With (Application) Holding a Standard Laser Button in The on Position During Experiments
For Use With (Equipment) Laser Diffraction Kit