Learn More
United Scientific™ Laser Line Generator
SDPSCI_ED
Catalog No. S000873 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Laser Line Generator
Catalog No. Product Type
S000873 Laser Line Generator
1 options
Prop 65
Catalog No. S000873 Supplier United Scientific™ Supplier No. RRT001L
Add to Cart
Add to Cart
Prop 65
The laser line generator is specially designed with a small glass rod in front of a Class 2 laser diode.
This refraction effect creates an easily visible line instead of a laser point. The line can be projected to a distance of over a meter. When this line is used with mirrors or lenses, the reflection and refraction paths are easy to see. For safety, the laser only stays on when the button is pressed. Device measures 2.75 x 1.25 x 1.25 in.
  • Visible line makes investigations into reflection and refraction a more vivid experience
  • Laser only stays on when the button is engaged
  • Safe Class 2 output (<1 mW)
  • Includes two AAA batteries
Warning: Always practice safe laboratory procedures when using a laser.
Specifications
Product Type Laser Line Generator