Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Laser Optical Disk Set
SDPSCI_ED

Catalog No. S139467 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S139467 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139467 Supplier United Scientific™ Supplier No. LODS01
Add to Cart
Add to Cart
This item is not returnable. View return policy

Set ideal for demonstrating ray optics.

This is an excellent and economical set for introductory ray optics.

  • The laser ray box projects one, three, or five parallel bright beams and is equipped with a momentary switch for safety.
  • A vinyl template with a degree circle and a linear scale forms the optical disk, and the set includes seven clear acrylic optical elements and a mirror with adjustable curvature for investigating refraction, lenses, prisms and mirrors.
  • The bottoms of the acrylic elements are frosted to show the ray paths inside them.
  • The set also includes a manual and storage box.
  • This product emits laser light. Max. output power < 1mW per beam. Wavelength 650 nm. Class IIIa laser product.
  • Includes one each of the following:
    • Wall Mount Power Supply
    • Laser Ray Box
    • Parallel sided Block
    • Trapezoidal Prism
    • Biconcave Lens
    • Biconvex Lens
    • Semicircular Lens (solid)
    • Semiciruclar Cell
    • Flexible template - ruler and protractor
    • Adjustable Mirror - plane/concave/convex
    • Aluminum Storage Case
    • Activity Guide

Specifications

Grade 5 to 12
Includes Wall-mount Power Supply, Laser Ray Box (1, 3, or 5 rays), Parallel-sided Block, Trapezoidal Prism, Right Angle Prism, Biconcave Lens, Biconvex Lens, Semicircular Lens (solid), Semicircular Cell, Flexible Template—ruler and protractor, Adjustable Mirror—plane/concave/convex, Aluminum Storage Case, Activity Guide
Class Size 1 Lab Group
Product Type Laser Optical Disk Set

Prop 65 Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.